missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPi2 Antibody (HIC1-2B4.2B), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-18946APCCY7
This item is not returnable.
View return policy
Description
HPi2 Monoclonal specifically detects HPi2 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| HPi2 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Tissue culture supernatant | |
| Primary | |
| Human, Mouse | |
| Supernatant |
| Flow Cytometry | |
| HIC1-2B4.2B | |
| Flow Cytometry | |
| human pancreatic endocrine islet cell | |
| Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
| 0.1 mL | |
| Specific for multiple endocrine cell types. | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction