missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPi3 Antibody (HIC1-7H10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-18947APC
This item is not returnable.
View return policy
Description
HPi3 Monoclonal antibody specifically detects HPi3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| HPi3 | |
| Monoclonal | |
| APC | |
| human pancreatic endocrine islet cell | |
| Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
| 0.1 mL | |
| Pancreatic Stem Cell Markers, Stem Cell Markers | |
| Human, Mouse | |
| Supernatant |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| HIC1-7H10 | |
| PBS | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction