missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPRG Antibody (28), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89677PE
This item is not returnable.
View return policy
Description
HPRG Monoclonal antibody specifically detects HPRG in Human samples. It is validated for ELISA
Specifications
| HPRG | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3273 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 28 | |
| ELISA | |
| DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human HPRG (Met 1-Lys 525; Accession#: NP_000403.1). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction