missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPRT1 Monoclonal Antibody (1F8D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515274
This item is not returnable.
View return policy
Description
MA5-15274 targets HPRT in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15274 immunogen is purified recombinant fragment of HPRT expressed in E. Coli.
The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.
Specifications
| HPRT1 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| P00492 | |
| HPRT1 | |
| Purified recombinant fragment of HPRT expressed in E. Coli. | |
| RUO | |
| 3251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1F8D11 | |
| Unconjugated | |
| HPRT1 | |
| C81579; HGPRT; HGPRTase; HPGRT; Hprt; HPRT 1; HPRT B; Hprt1; hprt1 protein; hypoxanthine guanine phosphoribosyl transferase; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; hypoxanthine phosphoribosyltransferase 1; hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; id:ibd1344; id:ibd5108; testicular tissue protein Li 89; wu:fc10g09; zgc:56221; zgc:86608 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction