Learn More
HPRT1 Mouse anti-Human, Clone: OTI1B1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF506952
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.Specifications
| HPRT1 | |
| Monoclonal | |
| Unconjugated | |
| HPRT1 | |
| HGPRT, HPRT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3251 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1B1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P00492 | |
| HPRT1 | |
| Full length human recombit protein of human HPRT1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.