missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31061
This item is not returnable.
View return policy
Description
HPS3 Polyclonal specifically detects HPS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HPS3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| HPS3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human HPS3. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.92mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| DKFZp686F0413, FLJ22704, Hermansky-Pudlak syndrome 3, Hermansky-Pudlak syndrome 3 protein, SUTAL | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84343 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction