missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HPS4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28905
This item is not returnable.
View return policy
Description
HPS4 Polyclonal antibody specifically detects HPS4 in Human samples. It is validated for Western Blot
Specifications
| HPS4 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| bK1048E9.4, Hermansky-Pudlak syndrome 4, Hermansky-Pudlak syndrome 4 protein, KIAA1667bK1048E9.5, LE, Light-ear protein homolog | |
| Between 658 and 708 | |
| 100 μg | |
| Signal Transduction | |
| 89781 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction