missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HPSE Polyclonal Antibody
GREENER_CHOICE

Product Code. 16642855 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16642855 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16642855 Supplier Invitrogen™ Supplier No. PA5106416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HPSE.

Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al. ). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPSE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HPSE
Gene Accession No. Q6YGZ1, Q9Y251
Gene Alias Endo-glucoronidase; endoglycosidase heparanase; HEP; Heparanase; Heparanase 50 kDa subunit; Heparanase 8 kDa subunit; Heparanase-1; heparanase-like; Hpa; HPA1; HPR1; HPSE; HPSE1; HSE1
Gene Symbols HPSE
Host Species Rabbit
Immunogen A synthesized peptide derived from human HPSE(Accession Q9Y251), corresponding to amino acid residues V323-A373.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10855, 15442
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.