missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 Antibody (HPV16/1295), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08758R
This item is not returnable.
View return policy
Description
HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HPV16 | |
| Monoclonal | |
| DyLight 550 | |
| Human Papilloma Virus Type 16 | |
| HPV-16 E6 peptide (MHQKRTAMFQDPPQERPRKLPQLC) | |
| 100 μg | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| HPV16/1295 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction