missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 Antibody (SPM405), Alexa Fluor™ 532 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11575AF532
This item is not returnable.
View return policy
Description
HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HPV16 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Human Papilloma Virus Type 16 | |
| Recombinant major capsid protein L1 from human papilloma virus type 16 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction