Learn More
HPV16 Antibody (SPM405), Janelia Fluor™ 646 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11575JF646
Product Code. 18597456
  • € 726.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
HPV16
Monoclonal
Janelia Fluor 646
Human Papilloma Virus Type 16
Recombinant major capsid protein L1 from human papilloma virus type 16
0.1 mL
Primary
Store at 4°C in the dark.
IgG2a κ
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
SPM405
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Virus