missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E2 Antibody (EL58), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50427AF350
This item is not returnable.
View return policy
Description
HPV16 E2 Monoclonal specifically detects HPV16 E2 in Virus samples. It is validated for Western Blot.
Specifications
| HPV16 E2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Cancer, Cell Cycle and Replication, Virology Bacteria and Parasites | |
| Virus | |
| IgG1 |
| Western Blot | |
| EL58 | |
| Western Blot | |
| HPV16 regulatory protein E2, Human Papilloma Virus 16 E2, Human papilloma virus 16 regulatory protein E2, Human papillomavirus type 16 E2, Human papillomavirus type 16 regulatory protein E2, Regulatory protein E2 | |
| Bacterial recombinant protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction