missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E2 Antibody (TVG 261), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2753FR
This item is not returnable.
View return policy
Description
HPV16 E2 Monoclonal antibody specifically detects HPV16 E2 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| HPV16 E2 | |
| Monoclonal | |
| DyLight 680 | |
| HPV16 regulatory protein E2, Human Papilloma Virus 16 E2, Human papilloma virus 16 regulatory protein E2, Human papillomavirus type 16 E2, Human papillomavirus type 16 regulatory protein E2, Regulatory protein E2 | |
| Vaccinia-E2 followed by intravenous injection of the maltose binding protein MBP-E2 (generated from an expression vector in which the E2 ORF was cloned in frame with malE). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| TVG 261 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction