missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E7 Antibody (3D6F1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51548-0.025ml
This item is not returnable.
View return policy
Description
HPV16 E7 Monoclonal specifically detects HPV16 E7 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Papillomavirus 16 E7 | |
| Monoclonal | |
| Unconjugated | |
| E7, HPV16 E7, HpV16gp2, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16 | |
| Purified full length E7 expressed in E. Coli. | |
| 0.025 mL | |
| Primary | |
| Virus | |
| Ascites |
| Western Blot | |
| 3D6F1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Human papillomavirus type 16. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction