missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E7 Antibody (TVG 701Y), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2768JF646
This item is not returnable.
View return policy
Description
HPV16 E7 Monoclonal specifically detects HPV16 E7 in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Papillomavirus16E7 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence | |
| TVG 701Y | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| E7, HPV16 E7, HpV16gp2, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16 | |
| HPV16 E7 synthesised from S. pombe | |
| 0.1 mL | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction