missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E6 Antibody (BF7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2769UV
This item is not returnable.
View return policy
Description
HPV18 E6 Monoclonal specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| HPV18 E6 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| BF7 | |
| Western Blot, ELISA | |
| E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 | |
| Gel-purified HPV18 E6-beta galactosidase fusion protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction