Learn More
HRAS Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99921APC
Product Code. 18681217
  • € 407.00 / 0.10mL
Description
HRAS Polyclonal antibody specifically detects HRAS in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HRAS
Polyclonal
C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1
3265
Purified
Western Blot
Western Blot, ELISA, Immunoprecipitation
Rabbit
Store at 4C in the dark.