missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody (OTI1D9), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74847AF405
This item is not returnable.
View return policy
Description
HRAS Monoclonal specifically detects HRAS in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| HRAS | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 3265.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HRAS | |
| Full length recombinant protein of human vH RAS (NP_005334) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction