missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody (OTI1D9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74847CL1
This item is not returnable.
View return policy
Description
HRAS Monoclonal antibody specifically detects HRAS in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| HRAS | |
| Monoclonal | |
| CoraFluor 1 | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| Full length human recombinant protein of human HRAS(NP_005334) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 3265 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1D9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction