missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRD1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2526MFV500
This item is not returnable.
View return policy
Description
HRD1 Polyclonal antibody specifically detects HRD1 in Human, Mouse, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
Specifications
| HRD1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated | |
| E3 ubiquitin-protein ligase synoviolin, EC 6.3.2, HRD1EC 6.3.2.-, KIAA1810Synovial apoptosis inhibitor 1, MGC40372, synovial apoptosis inhibitor 1, synoviolin | |
| A synthetic peptide made to an internal portion of human HRD1 (within residues 300-400). [UniProt Q96PK3] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Ubiquitin Proteasome Pathway | |
| 84447 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction