missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRG Monoclonal Antibody (C4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551002
This item is not returnable.
View return policy
Description
HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
Specifications
| HRG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q99PS8 | |
| HRG | |
| Recombinant Eukaryotic Rat HRG protein, Leu19-Lys525 (Accession #Q99PS8). | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C4 | |
| Unconjugated | |
| HRG | |
| AI265597; AW413091; D16jh2; D18020; DKFZp779H1622; histidine rich glycoprotein; histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; histidine-rich glycoprotein 1; HPRG; HRG; HRG1; Hrgp; THPH11 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 171016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction