missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRH2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142174
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
H2 Histamine Receptor stimulates adenylate cyclase activity and leads to the release of acids in the stomach. It is also involved in GI motility and intestinal secretion. Treatments for acid-peptic disorders are based on the inhibition of histamine H2 receptor activity. The 3-untranslated region may be involved in differential expression. Histamine H2 receptor has been reported to be expressed in bone marrow, brain, heart, lymph node, nose, placenta, spinal cord, and stomach. An EST has been isolated from a kidney library.
Specifications
| HRH2 | |
| Polyclonal | |
| Unconjugated | |
| HRH2 | |
| gastric receptor 1; gastric receptor I; H2R; HH2R; histamine H2 receptor; histamine receptor; histamine receptor H 2; histamine receptor H2; HRH2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3274 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P25021 | |
| HRH2 | |
| Peptide with sequence C-QEEKPLKLQVWSGTE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction