missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HRH3 Recombinant Rabbit Monoclonal Antibody (7H8E6)
GREENER_CHOICE

Product Code. 17206953 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17206953 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17206953 Supplier Invitrogen™ Supplier No. MA538074

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse testis, Mouse brain, Mouse spleen, Rat testis, Rat brain, Rat spleen, Raji, SH-SY5Y, U-251MG Immunogen sequence: MERAPPDGPL NASGALAGEA AAAGGARGFS AAWTAVLAAL MALLIVATVL GNALVMLAFV ADSSLRTQNN FFLLNLAISD FLVGAFCIPL YVPYVLTGRW.

H3 Histamine Receptor regulates the release of the neurotransmitters norepinephrine and somatostatin. In histamine-containing neurons of the brain, the H3 receptor acts as a presynaptic autoreceptor, while in nonhistamine-containing neurons, in both the central and peripheral nervous systems, it acts as a presynaptic heteroreceptor. Multiple isoforms of H3 receptors are produced by alternative splicing. Histamine H3 receptor has been reported primarily in brain. ESTs have been isolated from brain (normal and cancer), breast cancer, colon cancer, and retinoblastoma libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRH3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 7H8E6
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HRH3
Gene Accession No. P58406, Q9QYN8, Q9Y5N1
Gene Alias AW049250; G protein-coupled receptor; G protein-coupled receptor 97; GPCR97; G-protein coupled receptor 97; H3R; HH3R; histamine H3 receptor; histamine H3 receptor H3L isoform; histamine H3 receptor H3S isoform; histamine receptor; histamine receptor H 3; histamine receptor H3; Hrh3
Gene Symbols HRH3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HRH3 (Q9Y5N1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11255, 85268, 99296
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.