Learn More
Hrk Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76414MFV500
Product Code. 18455016
  • € 622.00 / 0.10mL
Description
Hrk Polyclonal antibody specifically detects Hrk in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hrk
Polyclonal
Western Blot, ELISA
activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5
Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Hrk. The immunogen is located within the last 50 amino acids of Hrk. Amino Acid Squence: MWRRRARSRRAPAPG
0.1 mL
Neuroscience, Signal Transduction
8739
Store at 4C in the dark.
IgG
Western Blot, ELISA
mFluor Violet 500 SE
50mM Sodium Borate
Rabbit
Peptide affinity purified