missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HROB Antibody (3E10H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | HROB |
|---|---|
| Clone | 3E10H2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18167687
|
Novus Biologicals
NBP2-37405 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610526
|
Novus Biologicals
NBP2-37405-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HROB Monoclonal specifically detects HROB in Human samples. It is validated for Western Blot, ELISA.Specifications
| HROB | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 78995 | |
| Purified recombinant fragment of human C17ORF53 (AA: 282-527) expressed in E. Coli. | |
| Primary | |
| 69.8 kDa |
| 3E10H2 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Human | |
| chromosome 17 open reading frame 53, FLJ11594, hypothetical protein LOC78995, MCM8IP, MGC3130 | |
| C17orf53 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title