missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRP-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05607-20ul
This item is not returnable.
View return policy
Description
HRP-2 Polyclonal antibody specifically detects HRP-2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| HRP-2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| HDGF Like 2, HDGF2, HDGF-2, HDGFL2, HDGF-Related Protein 2, HDGFRP2, Hepatoma-derived growth factor 2, Hepatoma-Derived Growth Factor Related Protein 2, Hepatoma-Derived Growth Factor-Related Protein 2, HRP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 501-670 of human HRP-2 (NP_116020.1). EELGTLQVTSQILQKNTDVVATLKKIRRYKANKDVMEKAAEVYTRLKSRVLGPKIEAVQKVNKAGMEKEKAEEKLAGEELAGEEAPQEKAEDKPSTDLSAPVNGEATSQKGESAEDKEHEEGRDSEEGPRCGSSEDLHDVREGPDLDRPGSDRQERERARGDSEALDEES | |
| 20 μg | |
| Cell Biology | |
| 84717 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction