missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRP-2 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32074V
This item is not returnable.
View return policy
Description
HRP-2 Polyclonal antibody specifically detects HRP-2 in Human samples. It is validated for Western Blot
Specifications
| HRP-2 | |
| Polyclonal | |
| Western Blot | |
| HDGF Like 2, HDGF2, HDGF-2, HDGFL2, HDGF-Related Protein 2, HDGFRP2, Hepatoma-derived growth factor 2, Hepatoma-Derived Growth Factor Related Protein 2, Hepatoma-Derived Growth Factor-Related Protein 2, HRP2 | |
| The immunogen this antibody was made to, maps to a region between residue 621 to 671 of human Hepatoma-Derived Growth Factor-Related Protein 2 using the numbering given in entry NP_001001520.1 (GeneID 84717). | |
| 0.1 mL | |
| Cell Biology | |
| 84717 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction