Learn More
HRP-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32074
Product Code. 18760914
  • € 570.00 / 100µL
Description
HRP-2 Polyclonal specifically detects HRP-2 in Human samples. It is validated for Western Blot, Immunoprecipitation.