missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRP Antibody (2H11cc), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-8035C
This item is not returnable.
View return policy
Description
HRP Monoclonal antibody specifically detects HRP in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry
Specifications
| HRP | |
| Monoclonal | |
| DyLight 650 | |
| EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified peroxidase from horseradish. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA, Immunohistochemistry | |
| 2H11cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction