missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRP Monoclonal Antibody (HP-03)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110371
This item is not returnable.
View return policy
Description
This antibody reacts with horseradish peroxidase. It is suitable for PAP detection system.
HRP, also known as horseradish peroxidase, is an enzyme that is commonly used in biochemistry and molecular biology research. It is derived from the root of the horseradish plant and catalyzes the oxidation of a wide range of substrates in the presence of hydrogen peroxide. HRP is widely used as a detection enzyme in immunoassays, such as ELISAs and Western blots, due to its high sensitivity and stability. It can also be used as a tracer in enzyme-linked immunosorbent assays (ELISAs) and as a labeling agent in fluorescent microscopy.
Specifications
| HRP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| Mouse | |
| Sequential chromatography | |
| RUO | |
| Chemical, Tag | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| HP-03 | |
| Unconjugated | |
| Horseradish Peroxidase | |
| Horseradish peroxidase (Grade I). | |
| 100 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid | |
| Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction