missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HRX Monoclonal Antibody (10F8D7)
GREENER_CHOICE

Product Code. 11335353
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335353 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11335353 Supplier Invitrogen™ Supplier No. MA515380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15380 targets MLL in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15380 immunogen is purified recombinant fragment of MLL (aa3751-3968) expressed in E. Coli.

KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRX
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10F8D7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KMT2A
Gene Accession No. Q03164
Gene Alias 6430520K01; ALL1; ALL-1; CDK6/MLL fusion protein; C-terminal cleavage product of 180 kDa; CXXC7; CXXC-type zinc finger protein 7; histone-lysine N-methyltransferase 2A; histone-lysine N-methyltransferase MLL; HRX; HTRX; HTRX1; Kmt2a; lysine (K)-specific methyltransferase 2A; lysine methyltransferase 2A; lysine N-methyltransferase 2A; mixed lineage leukemia 1; mKIAA4050; Mll; MLL cleavage product C180; MLL cleavage product N320; MLL/ENL fusion protein; MLL/GAS7; MLL/GAS7 fusion protein; MLL/GMPS fusion protein; MLL/hCDCrel fusion protein; MLL1; MLL1A; MLL-AF4 der(11) fusion protein; MLL-AF9; Myeloid/lymphoid or mixed-lineage leukemia; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia 1; myeloid/lymphoid or mixed-lineage leukemia protein 1; N-terminal cleavage product of 320 kDa; p180; p320; rearranged MLL protein; TET1-MLL; trithorax Drosophila; trithorax-like protein; TRX1; WDSTS; zinc finger protein HRX
Gene Symbols KMT2A
Host Species Mouse
Immunogen Purified recombinant fragment of MLL (aa3751-3968) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4297
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.