missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HS1BP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12845-1-AP-20UL
This item is not returnable.
View return policy
Description
Transcriptional activator binding to the E-box 1 core sequence of the E-cadherin promoter gene; the core-binding sequence is 5′CAGGTG-3′. Capable of reversing CTBP1-mediated transcription repression. Component of a splicing-dependent multiprotein exon junction complex (EJC) deposited at splice junction on mRNAs. The EJC is a dynamic structure consisting of a few core proteins and several more peripheral nuclear and cytoplasmic associated factors that join the complex only transiently either during EJC assembly or during subsequent mRNA metabolism. Participates in the regulation of alternative pre-mRNA splicing. Associates to spliced mRNA within 60 nt upstream of the 5′-splice sites. Involved in the establishment and maintece of epithelia cell-cell adhesion. Potential tumor suppressor for renal cell carcinoma.Specifications
| HS1BP3 | |
| Polyclonal | |
| Unconjugated | |
| HS1BP3 | |
| ETM2, HCLS1 binding protein 3, HS1 binding protein 3, HS1 BP3, HS1BP3, HSP1BP 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 313950, 58240, 64342 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TC93, Q53T59 | |
| Hs1bp3 | |
| HS1BP3 Fusion Protein Ag3880 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction