missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HS2ST1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589472

Product Code. 16507361

  • € 528.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MGLLRIMMPP KLQLLAVVAF AVAMLFLENQ IQKLEESRSK LERAIARHEV REIEQRHTMD GPRQDATLDE EEDMVIIYNR VPKTASTSFT NIAYDLCAKN KYHVLHINTT KNNPVMSLQD QVRFVKNITS WKEMKPGFYH GHVSYLDFAK FGVKKKPIYI NVIRDPIERL VSYYYFLRFG DDYRPGLRRR KQGDKKTFDE CVAEGGSDCA PEKLWLQIPF FCGHSSECW; Positive Samples: U-87MG, Mouse lung, Rat thymus; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. This gene encodes a member of the heparan sulfate biosynthetic enzyme family that transfers sulfate to the 2 position of the iduronic acid residue of heparan sulfate. The disruption of this gene resulted in no kidney formation in knockout embryonic mice, indicating that the absence of this enzyme may interfere with the signaling required for kidney formation. Two alternatively spliced transcript variants that encode different proteins have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HS2ST1
Polyclonal
Unconjugated
HS2ST1
2OST; 2-OST; 2-O-sulfotransferase; AW214369; dJ604K5.2; heparan sulfate 2-O-sulfotransferase 1; Hs2st; Hs2st1; KIAA0448; mKIAA0448
Rabbit
Affinity chromatography
RUO
23908, 292155, 9653
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.69 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q7LGA3, Q8R3H7
HS2ST1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-229 of human HS2ST1 (NP_0011279641).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.