missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HS3ST1 Polyclonal Antibody

Beskrivelse
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
HS3ST1 is the rate limiting enzyme for synthesis of HSact. HS3ST1 performs the crucial step modification in the biosynthesis of anticoagulant heparan sulfate (HSact) that is to complete the structure of the antithrombin pentasaccharide binding site. Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It possesses both heparan sulfate glucosaminyl 3-O-sulfotransferase activity, anticoagulant heparan sulfate conversion activity, and is a rate limiting enzyme for synthesis of anticoagulant heparan. This enzyme is an intraluminal Golgi resident protein.
Tekniske data
Tekniske data
| Antigen | HS3ST1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | HS3ST1 |
| Gene Accession No. | O14792 |
| Gene Alias | 3OST; 3-Ost; 3OST1; 3-OST-1; D5Wsu110e; h3-OST-1; heparan sulfate; heparan sulfate (glucosamine) 3-O-sulfotransferase 1; heparan sulfate 3-O-sulfotransferase 1; heparan sulfate 3-O-sulfotransferase-1; heparan sulfate 3-O-sulfotransferase-1 precursor; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 1; Heparan sulfate glucosamine 3-O-sulfotransferase 1; heparan sulfate-glucosamine 3-sulfotransferase 1; heparin-glucosamine 3-O-sulfotransferase; HS3ST1; Hsg3ost |
| Gene Symbols | HS3ST1 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?