missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hs6st3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111749
This item is not returnable.
View return policy
Description
Immunogen sequence: QQRYHHTKQL EHQRDRQKRR EERRLQREHR DHQWPKEDGA AEGTVTEDYN SQVVRW Highest antigen sequence indentity to the following orthologs: Rat - 43%, Mouse - 93%.
6-O-sulfation enzyme which catalyzes the transfer of sulfate from 3'-phosphoadenosine 5'-phosphosulfate (PAPS) to position 6 of the N-sulfoglucosamine residue (GlcNS) of heparan sulfate.
Specifications
| Hs6st3 | |
| Polyclonal | |
| Unconjugated | |
| Hs6st3 | |
| 6OST3; 6-ost-3; DKFZp761K2315; heparan sulfate 6-O-sulfotransferase 3; heparan sulfate 6-O-sulfotransferase 3b; heparan-sulfate 6-O-sulfotransferase 3; heparan-sulfate 6-O-sulfotransferase 3 pseudogene; heparan-sulfate 6-O-sulfotransferase 3-B; HS 6-OST-3-B; hs6st3; HS6ST-3; hs6st3b; RGD1560050; sb:eu962 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 266722 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZP7 | |
| Hs6st3 | |
| Recombinant Protein corresonding to Human Hs6st3. Recombinant protein control fragment (Product #RP-101543). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction