missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568701
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 86%.
Negative regulator of the heat shock response. Negatively affects HSF1 DNA-binding activity. May have a role in the suppression of the activation of the stress response during the aging process.
Specifications
| HSBP1 | |
| Polyclonal | |
| Unconjugated | |
| HSBP1 | |
| 0610007A03Rik; AA408769; heat shock factor binding protein 1; heat shock factor-binding protein 1; HSBP1; HSF1BP; Hsp25; Nasopharyngeal carcinoma-associated antigen 13; NPC-A-13; Unknown (protein for MGC:128054) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3281 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75506 | |
| HSBP1 | |
| synthetic peptide directed towards the N terminal region of human HSBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction