Learn More
HSD17B11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16847174 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16847174 20 μL 20µL
16837174 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16847174 Supplier Proteintech Supplier No. 173011AP20UL
Rabbit Polyclonal Antibody
Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.
Specifications
Antigen HSD17B11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSD17B11
Gene Accession No. Q6AYS8, Q8NBQ5, Q9EQ06
Gene Alias 17 beta HSD 11, 17 beta HSD XI, 17 BETA HSD11, 17 BETA HSDXI, 17betaHSD11, 17betaHSDXI, 17bHSD11, DHRS8, HSD17B11, PAN1B, RETSDR2, SDR16C2, UNQ207/PRO233
Gene Symbols Hsd17b11
Host Species Rabbit
Immunogen HSD17B11 Fusion Protein Ag11182
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114664, 289456, 51170
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG