missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B13 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33263-20ul
This item is not returnable.
View return policy
Description
HSD17B13 Monoclonal antibody specifically detects HSD17B13 in Human samples. It is validated for ELISA,Western Blot
Specifications
| HSD17B13 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| HMFN0376, hydroxysteroid (17-beta) dehydrogenase 13, member 3,17-beta-hydroxysteroid dehydrogenase 13, MGC138508,17-beta-HSD 13, NIIL497, SCDR9, short-chain dehydrogenase/reductase 9, UNQ497/PRO1014 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-211 of human HSD17B13 (NP_835236.2).,, Sequence:, RRKSVAGEIVLITGAGHGIGRQTTYEFAKRQSILVLWDINKRGVEETAAECRKLGVTAHAYVVDCSNREEIYRSLNQVKKEVGDVTIVVNNAGTVYPADLLSTKDEEITKTFEVNILGHFWITKALLPSMMERNHGHIVTVASVCGHEGIPYLIPYCSSKFAAVGFHRGLTSELQALGKTGI | |
| 20 μL | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 345275 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction