missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B2 Antibody (OTI4F10), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70945DL594
This item is not returnable.
View return policy
Description
HSD17B2 Monoclonal antibody specifically detects HSD17B2 in Human samples. It is validated for Simple Western, Flow Cytometry
Specifications
| HSD17B2 | |
| Monoclonal | |
| DyLight 594 | |
| 17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, E2DH, EC 1.1.1.62,17-beta-HSD 2, EC 1.1.1.63, EDH17B220-alpha-HSD, estradiol 17-beta-dehydrogenase 2, HSD17, hydroxysteroid (17-beta) dehydrogenase 2, Microsomal 17-beta-hydroxysteroid dehydrogenase, SDR9C2, short chain dehydrogenase/reductase family 9C, member 2, Testosterone 17-beta-dehydrogenase | |
| Full length recombinant protein of human HSD17B2 (NP_002144) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Simple Western, Flow Cytometry | |
| OTI4F10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 3294 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction