missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B2 Monoclonal Antibody (OTI4F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504565S
This item is not returnable.
View return policy
Description
HSD17B2 Monoclonal antibody specifically detects HSD17B2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HSD17B2 | |
| Monoclonal | |
| 0.45 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHQ9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51706 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4F10 | |
| Unconjugated | |
| CYB5R1 | |
| 1.1.1.239; 17 beta HSD 2; 17 HSD type 2; 17-beta hydroxysteroid dehydrogenase type 2; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha HSD; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; AI194836; AI194967; AI255511; E2DH; EDH17B2; estradiol 17-beta-dehydrogenase 2; HSD17; HSD17B2; hydroxysteroid (17-beta) dehydrogenase 2; hydroxysteroid 17-beta dehydrogenase 2; microsomal 17-beta-hydroxysteroid dehydrogenase; SDR9C2; short chain dehydrogenase/reductase family 9C member 2; short chain dehydrogenase/reductase family 9C, member 2; tes; testosterone 17-beta-dehydrogenase | |
| Full length human recombinant protein of human CYB5R1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction