missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B4 Antibody (OTI4C4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70947B
This item is not returnable.
View return policy
Description
HSD17B4 Monoclonal specifically detects HSD17B4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| HSD17B4 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| HSD17B4 | |
| Full length recombinant protein of human HSD17B4 (NP_000405) produced in HEK293T cells. | |
| 0.1 mL | |
| 3295.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4C4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase, 7-alpha, D-bifunctional protein, EDH17B4, hydroxysteroid (17-beta) dehydrogenase 4, MPF-2, peroxisomal, peroxisomal multifunctional enzyme type 2,17-beta-HSD IV, peroxisomal multifunctional protein 2,17beta-estradiol dehydrogenase type IV, short chain dehydrogenase/reductase family 8C, member 1,3-alpha | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction