missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B4 Mouse anti-Human, Clone: OTI2C9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507075S
This item is not returnable.
View return policy
Description
HSD17B4 Monoclonal antibody specifically detects HSD17B4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| HSD17B4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P51659 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3295 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C9 | |
| Unconjugated | |
| HSD17B4 | |
| (3R)-hydroxyacyl-CoA dehydrogenase; 17[b]-HSD; 17beta-estradiol dehydrogenase type IV; 17-beta-HSD; 17-beta-HSD 4; 17-beta-HSD IV; 17-beta-hydroxysteroid dehydrogenase 4; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase; 4; beta-hydroxyacyl dehydrogenase; beta-keto-reductase; D-3-hydroxyacyl-CoA dehydratase; D-bifunctional protein; D-bifunctional protein, peroxisomal; DBP; Edh17b4; Enoyl-CoA hydratase 2; Hsd17b4; hydroxysteroid (17-beta) dehydrogenase 4; hydroxysteroid 17-beta dehydrogenase 4; hydroxysteroid dehydrogenase 4; MFE-2; MFP2; Mfp-2; MPF-2; Multifunctional protein 2; perMFE-2; peroxisomal multifunctional enzyme type 2; peroxisomal multifunctional enzyme type II; peroxisomal multifunctional protein 2; PRLTS1; SDR8C1; Short chain dehydrogenase/reductase family 8C member 1; short chain dehydrogenase/reductase family 8C, member 1 | |
| Full length human recombinant protein of human HSD17B4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction