missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B7, Mouse anti-Human, Clone: 1G10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00051478-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: NAFTLTPYNGTEALVWLFHQKPESLNPLIKYLSATTGFGRNYIMTQKMDLDEDTAEKFYQKLLELEKHIRVTIQKTDNQARLSGSCLSpecifications
| HSD17B7 | |
| Monoclonal | |
| Unconjugated | |
| hydroxysteroid (17-beta) dehydrogenase 7 | |
| MGC12523/MGC75018/PRAP/SDR37C1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1G10 | |
| In 1x PBS, pH 7.4 | |
| NM_016371 | |
| HSD17B7 | |
| HSD17B7 (NP_057455.1, 255 a.a. ∼ 341 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51478 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction