missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSD17B7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17731934
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17731934 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17731934 Supplier Invitrogen™ Supplier No. PA5120533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse liver, Mouse ovary. The target is usually found in the following locations: Cell membrane, Single-pass membrane protein. Immunogen sequence: SRKVIHMFST AEGLLTQGDK ITADGLQEVF ETNVFGHFIL IRELEPLLCH SDNPSQLIWT SSRSARKSNF SLEDFQHSKG KEPYSSSKYA T.

Responsible for the reduction of the keto group on the C-3 of sterols.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HSD17B7
Gene Accession No. O88736, P56937
Gene Alias 17 beta-hydroxysteroid dehydrogenase type VII; 17beta hydroxysteroid dehydrogenase; 17-beta-HSD 7; 17-beta-hydroxysteroid dehydrogenase 7; 3-keto-steroid reductase; AI266814; bA291L22.1; ERG27; Estradiol 17-beta-dehydrogenase 7; Hsd17b_2; HSD17B7; HSD17B7P2; hydroxysteroid (17-beta) dehydrogenase 7; hydroxysteroid 17-beta dehydrogenase 7; hydroxysteroid 17-beta dehydrogenase 7 pseudogene 2; hydroxysteroid dehydrogenase 17 beta, type 7; PRAP; PRL receptor-associated protein; SDR37C1; Short chain dehydrogenase/reductase family 37C member 1; short chain dehydrogenase/reductase family 37C, member 1; UNQ2563/PRO6243
Gene Symbols HSD17B7
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15490, 51478
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.