missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B8 Antibody (OTI2E12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71350FR
This item is not returnable.
View return policy
Description
HSD17B8 Monoclonal specifically detects HSD17B8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HSD17B8 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| HSD17B8 | |
| Recombinant protein fragment corresponding to amino acids 57-261 of human HSD17B8 (NP_055049) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2E12 | |
| Western Blot, Immunohistochemistry | |
| D6S2245EdJ1033B10.9, EC 1.1.1.-, EC 1.1.1.62, EC 1.1.1.63,17-beta-HSD 8, estrogen 17-oxidoreductase, FABGLestradiol 17-beta-dehydrogenase 8, H2-KE6, HKE6FabG (beta-ketoacyl-[acyl-carrier-protein] reductase, E coli) like (E. coli), hydroxysteroid (17-beta) dehydrogenase 8,3-oxoacyl-[acyl-carrier-protein] reductase, ke-6, KE6, Ke-6,17-beta-hydroxysteroid dehydrogenase 8, Protein Ke6, Really interesting new gene 2 protein, RING2FABG, SDR30C1, short chain dehydrogenase/reductase family 30C, member 1, Testosterone 17-beta-dehydrogenase 8 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 7923.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction