missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B8 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800478S
This item is not returnable.
View return policy
Description
HSD17B8 Monoclonal antibody specifically detects HSD17B8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| HSD17B8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43366 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4112 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2E12 | |
| Unconjugated | |
| MAGEB1 | |
| 17-beta-HSD 8; 17-beta-hydroxysteroid dehydrogenase 8; 3-ketoacyl-[acyl-carrier-protein] reductase alpha subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; D17H6S112E; D6S2245E; dJ1033B10.9; Estradiol 17-beta-dehydrogenase 8; estrogen 17-oxidoreductase; FABG; FABGL; H FABGL; H2-K region expressed gene 6; H-2Ke6; H2-KE6; Hke6; Hsd17b8; hydroxysteroid (17-beta) dehydrogenase 8; hydroxysteroid 17-beta dehydrogenase 8; KAR alpha subunit; KE6; Ke-6; Protein Ke6; really interesting new gene 2 protein; RING2; SDR30C1; Short chain dehydrogenase/reductase family 30C member 1; short chain dehydrogenase/reductase family 30C, member 1; Testosterone 17-beta-dehydrogenase 8 | |
| Full length human recombinant protein of human MAGEB1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction