missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B8 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800478
This item is not returnable.
View return policy
Description
NAD-dependent 17-beta-hydroxysteroid dehydrogenase with highest activity towards estradiol. Has very low activity towards testosterone. The heteroteramer with CBR4 has NADH-dependent 3- ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria.Specifications
| HSD17B8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92506 | |
| HSD17B8 | |
| Human recombit protein fragment corresponding to amino acids 57-261 of human HSD17B8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2E12 | |
| Unconjugated | |
| HSD17B8 | |
| D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7923 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction