missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD3B7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101352
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HSD3B7.
This gene encodes an enzyme which is involved in the initial stages of the synthesis of bile acids from cholesterol and a member of the short-chain dehydrogenase/reductase superfamily. The encoded protein is a membrane-associated endoplasmic reticulum protein which is active against 7-alpha hydrosylated sterol substrates. Mutations in this gene are associated with a congenital bile acid synthesis defect which leads to neonatal cholestasis, a form of progressive liver disease. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Dec 2008].
Specifications
| HSD3B7 | |
| Polyclonal | |
| Unconjugated | |
| HSD3B7 | |
| 3 beta-hydroxy-delta 5-C27-steroid oxidoreductase; 3 beta-hydroxysteroid dehydrogenase type 7; 3 beta-hydroxysteroid dehydrogenase type VII; 3-beta-HSD VII; 3-beta-hydroxy-Delta(5)-C27 steroid oxidoreductase; AI195443; BB098564; C(27) 3-beta-HSD; C(27)-3BETA-HSD; CBAS1; Cca2; cholest-5-ene-3-be; cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase; Confluent 3Y1 cell-associated 2; HSD3B7; hsd3b7 {ECO:0000312; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7; PFIC4; RGD:628727}; SDR11E3; short chain dehydrogenase/reductase family 11E, member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 101502, 246211, 80270 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35048, Q9EQC1, Q9H2F3 | |
| HSD3B7 | |
| A synthesized peptide derived from human HSD3B7(Accession Q9H2F3), corresponding to amino acid residues R122-E172. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction