missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Antibody (10H4), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-42205C
This item is not returnable.
View return policy
Description
HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| HSF1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HSF1 | |
| Purified recombinant mouse HSF1 protein | |
| 0.1 mL | |
| Primary | |
| Detects approx 85kDa (unstressed cell lysates) and approx 95kDa (heat shocked cell lysates). | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 10H4 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| Rat | |
| Protein G purified | |
| Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
| 3297 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction