missing translation for 'onlineSavingsMsg'
Learn More

HSF1 Antibody (10H8), Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97504-0.025MG

 View more versions of this product

Product Code. 18080149

  • € 175.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSF1
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
HSF1
Recombinant mouse HSF1.
0.025 mg
Primary
Rabbit, Rat, Primate, Human, Mouse
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
10H8
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Rat
Protein G purified
RUO
3297
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado

For Research Use Only